Soft Care® Impact Foam Instant Hand Sanitizer 
Supplier: DIVERSEY INC.
100907873
76547-780CT 213.71 USD
76547-780
Soft Care® Impact Foam Instant Hand Sanitizer 
Skincare Soaps and Skin Cleansers
Soft Care® ImpactTM/MC is an alcohol based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protect against irritation for even the most frequent users.
  • Contains 76% (v/v), 72% (w/w) ethyl alcohol, meeting APIC, PHAC, CDC and WHO Healthcare Personal Hand Wash guidelines
  • Skin softening emollients leave hands soft and with no sticky residue
  • Made with 96% biobased ingredients
  • Dye-free and fragrance-free formulas reduce potential risk of skin irritation and allergic reactions for users with sensitive skin
  • NSF E-3 certified for use in food handling establishments
  • Minimize the risk of sick leave resulting from cross contamination
Fragrance-free and dye-free.
Soft Care® ImpactTM/MC meets CDC, WHO, APIC and PHAC hand hygiene recommendations for Healthcare personnel hand rubs.