Divoflow TC86, Low Foam Chlorinated Detergent for Hard Water
Supplier: DIVERSEY INC.
57760010
76547-754EA 1114.06 USD
76547-754
Divoflow TC86, Low Foam Chlorinated Detergent for Hard Water
Cleaning Agents Detergents and Surface Cleansers
Divoflow TC 86 cleaner is a low foaming, chlorinated alkaline detergent for CIP and spray washing use in the food, beverage and dairy industries.
  • Highly efficient in the removal of organic material including food soil, stains and off-flavors
  • Low foaming under conditions of high turbulence
  • Good hard water control and free rinsing, leaves no hard water films, minimizes acid cleaning cycles in most clean-in-place (CIP) operations
  • Free-rinsing, minimizes water usage
  • Liquid can be fed and controlled automatically in CIP systems