Acifoam VF10, Acidic Foam Cleaner-Descaler
Supplier: DIVERSEY INC.
57746100
76547-752EA 286.29 USD
76547-752
Acifoam VF10, Acidic Foam Cleaner-Descaler
Cleaning Agents Detergents and Surface Cleansers
Acifoam cleaner is based on inorganic acid and a blend of high foaming wetting agents/emulsifiers
  • Effective scale remover.
  • Versatile - Suitable for use on aluminum
  • Easy to use & Free-rinsing
  • Foam application extends contact time with vertical surfaces
It is specifically formulated for the removal of water hardness scale and other mineral deposits.
This product is also recommended for the periodic cleaning and descaling of high temperature processing equipment, such as cooking vessels, and also the exteriors of filler equipment, storage tanks and conveyers.
It is also effective for daily application to break down protein deposits in the fishing industry.
This product is suitable for use with a wide range of foam application equipment