Alpha-HP® Multi-Surface Disinfectant Cleaner
Supplier: DIVERSEY INC.
5549271
76547-702CT 132.81 USD
76547-702
Alpha-HP® Multi-Surface Disinfectant Cleaner
Cleaning Agents Disinfectants
An all-in-one, multi-surface disinfectant cleaner based on proprietary Accelerated Hydrogen Peroxide (AHP®) technology.
  • Bactericidal and virucidal, non-food contact sanitizer
  • Hydrogen peroxide active ingredient breaks down to oxygen and water after use
  • Excellent cleaning performance and environmental profile
  • Meets daily cleaning and disinfection needs
One product cleans and brightens surfaces safely and easily.
Use on walls, countertops and most other water washable hard surfaces.