Easy Paks® Detergent and Disinfectant
Supplier: DIVERSEY INC.
5412135
76547-698EA 83.85 USD
76547-698
Easy Paks® Detergent and Disinfectant
Cleaning Agents Disinfectants
Easy Paks® Detergent and Disinfectant is a concentrated powdered compound that quickly dissolves in water to produce a cleaning disinfectant dilution to meet your needs. 
  • Effectively cleans, disinfects, and deodorizes in one cleaning step
  • Effective against bacteria and viruses including HIV-1 (AIDS virus) and controls mildew
  • The effervescing action guarantees that both the cleaner and the packet will dissolve, quickly and completely
  • There's no guesswork, waste or messy clean up
  • The handy storage tub saves space and weighs less than 3 pounds
Cleans, deodorizes and disinfects hard, nonporous surfaces.
Effective against bacteria and viruses including HIV-1 (AIDS virus) and controls mildew.
Ideal for use in food service, nursing homes and other settings where housekeeping and disinfection are important.