Morning Mist™ Neutral Disinfectant Cleaner #33
Supplier: DIVERSEY INC.
5283038
76547-696CT 124.05 USD
76547-696
Morning Mist™ Neutral Disinfectant Cleaner #33
Cleaning Agents Disinfectants
A quaternary-based non-alkaline disinfectant with excellent cleaning properties.
  • Disinfectant cleaner that is bactericidal, virucidal and also kills MRSA and VRE
  • Hospital use disinfectant with a 10 minute contact time that meets bloodborne pathogen standards for decontaminating blood and bodily fluids
  • General cleaner and disinfectant for larger areas such as operating rooms, and patient care facilities.
Can be used in healthcare, hotels, schools and restaurants.
10-minute contact time. Bactericide and virucide. Kills MRSA, and VRE.
Meets bloodborne pathogen standards for decontaminating blood and body fluids.
Blue-green in color with a fresh scent.