Liquid Petroleum Gas Hydrometer, Specific Gravity with Thermometer
Supplier: Heathrow Scientific
67101HTS
76546-548EA 65.44 USD
76546-548
Liquid Petroleum Gas Hydrometer, Specific Gravity with Thermometer
Hydrometers
These liquid petroleum gas (LPG) hydrometer measures the specific gravity of LPG such as propane, butane, and other light hydrocarbons.
  • Calibrated for use at 15 °C (60 °F)
  • Thermometer contains safety blue liquid fill
  • Specific gravity range 0.500 to 0.650
It includes an internal thermometer for temperature measurement.