Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
Supplier: Heathrow Scientific
6613-B-C 6613-F-C 6613-G-C 6613-H-C 6613-I-C 6613-K-C 6613-L-C 6613-M-C 6613-N-C 6613-P-C 6613-Q-C 6613-R-C
76546-484EA 390.64 USD
76546-484 76546-488 76546-492 76546-496 76546-500 76546-504 76546-508 76546-512 76546-516 76546-520 76546-524 76546-528
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
Hydrometers
These IRS alcohol hydrometers use the Proof scale and comply with IRS specifications.
  • Calibration for use at 15.5 °C/60 °F
  • NIST-Traceable certificate
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
Delivery information: Includes hydrometer, NIST-traceable certificate, and PVC case.