VEE GEE Serological Pipette, Wide Tip
Supplier: Heathrow Scientific
20200-101A
76546-036CS 5617.46 USD
76546-036
VEE GEE Serological Pipette, Wide Tip
Pipettes Serological Pipettes
Our wide tip serological pipettes are meticulously manufactured for tolerances and accuracy in order to ensure pinpoint precision for all of your liquid handling needs.
  • Made out of high-quality borosilicate glass
  • Wide range of sizes to choose from
  • Reusable
  • Color-coded with permanent enamel markings
  • Calibrated to deliver
These serological pipettes are made out of high-quality borosilicate glass and feature a wide tip opening that facilitates a rapid fill and drain of viscous liquids. These pipettes also have graduation markings and color-coded bands that are permanently enameled to the glass which allows for quick identification of different volume ranges.
Certifications: Meets ASTM specifications and standards.
Delivery information: Comes in packs of 12.