Vee Gee Filtering Flasks
Supplier: Heathrow Scientific
1780T-1000 1780T-2000 1780T-300 1780T-500
76546-018EA 41.46 USD
76546-018 76546-020 76546-022 76546-024
Vee Gee Filtering Flasks
Flasks Filter Flasks
Filtering flasks are constructed of heavy wall borosilicate glass in order to withstand high vacuum applications.
  • Made of premium borosilicate glass
  • Heavy-duty rims to reduce chipping
  • Permanent white graduations
  • Large marking square for labeling
  • Port for tubulation has an outside diameter of 10 mm
  • Accepts a No. 8 rubber stopper (Not included)
Used in conjunction with vacuum filter systems, these filtering flasks feature heavy-duty rims to reduce chipping. The flasks also feature permanent white graduations and a large marking square for labeling. All flasks are sold individually.