EVE™ HT Automated Cell Counter, NanoEntek
Supplier: NANOENTEK USA INC.
EVE-HT
76545-832EA 48424.47 USD
76545-832
EVE™ HT Automated Cell Counter, NanoEntek
Cell Counters and Flow Cytometers Electronic Cell Counters
The EVE™ HT is a high-throughput automated cell counter, that can count 48 samples in just 3 minutes. Optimized with trypan blue staining, the system will discriminate live from dead cells precisely to provide reliable data on cell viability. With small sample volume and extensive capacity, the EVE™ HT provides accurate cell counting and viability measurements.
  • Disposable counting plates improve lab safety
  • Cell type: Cell lines
  • 10 μl required for each sample
  • User-independent consistency in results
  • Cell declustering technique to individually count cells
  • Exclude debris from results
  • Operation system: Windows 10 and 11
The 48-well counting plate designed for EVE™ HT can be used with a multi-pipette when mixing and loading the samples for high efficiency. Also, EVE™ HT comes with a very intuitive & powerful software that brings high productivity to the users. It is a perfect solution for cell line development and large scale cell production.
EVE™ HT offers an optional feature to safeguard data integrity required by 21 CFR Part 11. With this feature, it can not only manage users and give authority to specific users to manage data, but also save every user activity and create an audit trail.
Equipped with advanced optics and image analysis algorithms for counting , EVE™ HT can identify clumped cell populations at a precision that is proven to be comparable to more complex and fluorescence-based counting methods.