PMBBB Parallel Artificial Membrane Permeability Assay-Bbb Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
PMBBB-096
76545-502KT 484.67 USD
76545-502
PMBBB Parallel Artificial Membrane Permeability Assay-Bbb Kit, BioAssay Systems
Assays
Assess blood brain barrier (BBB) membrane permeability of test compounds.
  • Convenient: Includes all necessary equipment to run a PAMPA plate
  • Simple and low-cost: Procedure is easy to follow and more affordable than cell-based permeability assays
  • High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: PAMPA
  • Samples: Chemical compounds
MEMBRANE PERMEABILITY is an important characteristic to determine for evaluating compounds as potential drug candidates. Drugs often need to cross cell membranes in order to reach their target of action and this makes a compound’s ability to passively cross these membranes an important characteristic to evaluate. The Blood Brain Barrier (BBB) is made of brain endothelial cells with tight junctions. Rapid and early screening of compounds for BBB penetration is highly desirable for drug discovery. Permeability can be evaluated by cell-based methods; however, these methods are often expensive and time consuming.
Parallel Artificial Permeability Assays (PAMPA) offer researchers a quick, inexpensive method of evaluating the permeability of test compounds. Our PMBBB-096 kit is designed to aid in evaluating BBB permeability. BioAssay Systems' PMBBB Kit provides all the necessary components to run a Parallel Artificial Permeability Assay for Blood Brain Barrier studies.