QuantiChrom™ HRP Detection Reagent, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
DTMB-200
76545-500KT 190.7 USD
76545-500
QuantiChrom™ HRP Detection Reagent, BioAssay Systems
Assays ELISAs
Direct assays of peroxidases, ELISA, immunochromogenic chemistry, western blot.
  • Method: OD 450 nm
  • Samples: Peroxidases and HRP in ELISA
  • Species: All
  • Detection limit: 76 nU/well, or 7.7 pg/well
HORSERADISH PEROXIDASE (HRP) is an enzyme commonly used when conjugated to analytes of interest. When combined with hydrogen peroxide and a dye, HRP can be used to colorimetrically quantify the analyte.