CBS Cryogenic Freezers, S Series, Biolife Solutions
Supplier: CUSTOM BIOGENIC SYSTEMS, INC
33305
76542-168EA 33840.15 USD
76542-168
CBS Cryogenic Freezers, S Series, Biolife Solutions
Cryopreservation Systems
The S-3000AB standard LN₂ freezer has a stainless-steel insulated vessel, it has a round, space-saving design and is caster mounted for easy mobility. A counter-balanced lockable lid with a wide opening ensures easy, unrestricted access to samples. The unit has an advanced woven lid gasket design. Units provide flexibility to operate vapor storage, immersion storage, or a combination of both to store any type or size of the sample and feature annular fill and sensor tubes.
  • Provides long-term sample storage solution
  • The wide lid opening gives you quick access to your samples
  • Quick access without removing racks to find the one you need
  • Temperature uniformity
  • Temperatures up to -193 ⁰C
Model S-3000AB standard LN₂ container includes series 2301 Autofill-Monitor system which has autofill functionality with gas by-pass and LN₂ hose, dual temperature probes, and many additional features.