CBS 2101 Controlled Rate Freezer Cart, Biolife Solutions
Supplier: CUSTOM BIOGENIC SYSTEMS, INC
3120-F601
76542-166EA 3098.72 USD
76542-166
CBS 2101 Controlled Rate Freezer Cart, Biolife Solutions
Carts
Roller cart makes it easy to move the freezer in the laboratory.
  • The cart is designed to mount the 2101 CRF meaning the freezer can easily be moved where and when it is required
  • Shelves underneath the freezer provide storage or working space for racks and other laboratory tools
Optional roller cart for 2101 Controlled Rate Freezing system.