Xylene (mixture of isomers) ≥99.50%, Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger

Synonyms: Dimethylbenzene

XYLENELM-GAL XYLENELM-QT
76541-004CS 166.73 USD
76541-004 76541-064
Xylene (mixture of isomers) ≥99.50%, Low-Metal (LM) grade semiconductor grade
Xylene (mixture of isomers)

Transene Xylenes solvent is a "low metals" chemical suitable for demanding semiconeuctor and elecrtronics processing.

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –25 °C
Density: 0.85 g/cc
Flash Pt: 29 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3 II
Merck Index: 13,10136

Order Now

Specification Test Results

Xylene 99.5 min % Weight
Color 10 APHA max.
Acidity 0.3 ueq/g max.
Residue after Evaporation 5000 ppb (umg/g) max.
Moisture 0.02 % Weight max.
Chloride 3000 ppb (umg/g) max.
Phosphate 1000 ppb (umg/g) max.
Sulfur Compounds 30000 ppb (umg/g) max.
IR Spectra ACS Pass
Appearance Clear Liquid
Sub. Dark. BY H2SO4 Pass
Density (at 25°C) To Report g/mL
Aluminum 20 ppb (umg/g) max.
Antimony 10 ppb (umg/g) max.
Arsenic 10 ppb (umg/g) max.
Barium 20 ppb (umg/g) max.
Boron 20 ppb (umg/g) max.
Cadmium 20 ppb (umg/g) max.
Calcium 20 ppb (umg/g) max.
Chromium 20 ppb (umg/g) max.
Cobalt 20 ppb (umg/g) max.
Copper 10 ppb (umg/g) max.
Gallium 20 ppb (umg/g) max.
Gold 20 ppb (umg/g) max.
Iron 20 ppb (umg/g) max.
Lead 20 ppb (umg/g) max.
Lithium 20 ppb (umg/g) max.
Magnesium 20 ppb (umg/g) max.
Manganese 20 ppb (umg/g) max.
Molybdenum 20 ppb (umg/g) max.
Nickel 10 ppb (umg/g) max.
Potassium 20 ppb (umg/g) max.
Silicon 20 ppb (umg/g) max.
Silver 20 ppb (umg/g) max.
Sodium 20 ppb (umg/g) max.
Tin 20 ppb (umg/g) max.
Titanium 20 ppb (umg/g) max.
Zinc 20 ppb (umg/g) max.
Particle >1.0 μm 10 pcs/mL max.
Particle >0.5 μm 150 pcs/mL max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR