Ammonia solution 29%, Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

AMMYDLM-GAL AMMYDLM-QT
76541-026CS 144.78 USD
76541-026 76541-060
Ammonia solution 29%, Low-Metal (LM) grade semiconductor grade
Ammonia solution

Transene Ammonium Hydroxide 29% LM is a "low metals" grade suitable for semiconductor and electronics processing.

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 36 °C (H2O)
Melting Pt: –72 °C
Density: 0.9 g/cc
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Assay (NH3) 30 % max.
Density 0.91, nominal g/cc
Color 10 max.
Appearance Clear, sediment-free
Reducing Substance as KMnO4 P/F ppm
Carbon Dioxide (CO2) 20 ppm max.
Residue on Ignition 20 ppm max.
Non-Volatile Residue 5 mg/100 mL max.
Aluminum (Al) 10 ppb max.
Barium (Ba) 10 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 10 ppb max.
Boron (B) 10 ppb max.
Cadmium (Cd) 10 ppb max.
Calcium (Ca) 10 ppb max.
Chloride (Cl) 0.2 ppm max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 10 ppb max.
Heavy Metals (as As) 5 ppb max.
Iron (Fe) 10 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Niobium (Nb) 10 ppb max.
Nitrate (NO3) 0.4 ppm max.
Phosphate (PO4) 0.1 ppm max.
Potassium (K) 10 ppb max.
Silicon (Si) 10 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 10 ppb max.
Strontium (Sr) 10 ppb max.
Sulfate (SO4) 0.2 ppm max.
Tantalum (Ta) 10 ppb max.
Thallium (Tl) 10 ppb max.
Tin (Sn) 10 ppb max.
Titanium (Ti) 10 ppb max.
Vanadium (V) 10 ppb max.
Zinc (Zn) 10 ppb max.
Zirconium (Zr) 10 ppb max.
Particle Count, 0.5 um 150 ct/ml max.
Particle Count, 1.0 um 25 ct/ml max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR