Hydrogen peroxide 30%, Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger

060-H2O230LM-GAL 060-H2O230LM-QT
76540-940CS 133 USD
76540-940 76541-046
Hydrogen peroxide 30%, Low-Metal (LM) grade semiconductor grade
Hydrogen peroxide

Transene Hydrogen Peroxide LM is a "low metals" peroxide solution suitable for demanding semiconductor and electronics processing. Stabilizer is not needed at these low metal impurity levels.

Formula: H₂O₂
MW: 34.01
Boiling Pt: 150.2
Melting Pt: -0.41
Density: 1.11 g/cc
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1, 8 II
Merck Index: 14,04798

Order Now

Specification Test Results

Assay (H2O2) 30 - 32 %
Acidity (As H2SO4) 0.04 μeq/g max.
Color (APHA) 6 APHA
TMA (Trimethylamine) 2 ppb max.
Density @25 oC) 1.11 g/ml max.
Total Carbon (TC) 20000 ppb max.
Residue after Evaporation (RAE) 5000 ppb max.
Heavy Metals (HM) 50 ppb max.
Aluminum (Al) 1.0 ppb max.
Ammonium (NH4) 2000 ppb max.
Arsenic and Antimony (as As) 1.0 ppb max.
Barium (Ba) 1.0 ppb max.
Beryllium (Be) 1.0 ppb max.
Bismuth (Bi) 1.0 ppb max.
Boron (B) 1.0 ppb max.
Cadmium (Cd) 1.0 ppb max.
Calcium (Ca) 1.0 ppb max.
Chloride (Cl) 30 ppb max.
Chromium (Cr) 1.0 ppb max.
Cobalt (Co) 1.0 ppb max.
Copper (Cu) 1.0 ppb max.
Gallium (Ga) 1.0 ppb max.
Germanium (Ge) 1.0 ppb max.
Gold (Au) 1.0 ppb max.
Iron (Fe) 1.0 ppb max.
Lead (Pb) 1.0 ppb max.
Lithium (Li) 1.0 ppb max.
Magnesium (Mg) 1.0 ppb max.
Manganese (Mn) 1.0 ppb max.
Molybdenum (Mo) 1.0 ppb max.
Nickel (Ni) 1.0 ppb max.
Nitrate (NO3) 300 ppb max.
Phosphate (PO4) 100 ppb max.
Platinum (Pt) 1.0 ppb max.
Potassium (K) 1.0 ppb max.
Silicon (Si) 10 ppb max.
Silver (Ag) 1.0 ppb max.
Sodium (Na) 1.0 ppb max.
Strontium (Sr) 1.0 ppb max.
Sulfate (SO4) 100 ppb max.
Tantalum (Ta) 1.0 ppb max.
Thallium (Tl) 1.0 ppb max.
Tin (Sn) 1.0 ppb max.
Titanium (Ti) 1.0 ppb max.
Vanadium (V) 1.0 ppb max.
Zinc (Zn) 1.0 ppb max.
Zirconium (Zr) 1.0 ppb max.
Particle Count, 0.2 μm 500 ct/ml max.
Particle Count, 0.5 μm 100 ct/ml max.
Particle Count, 1.0 μm 25 ct/ml max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR