Acetic acid ≥99.70%, Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

060-3550-00-GL 060-3550-00-QT
76541-034CS 197.37 USD
76541-034 76541-044
Acetic acid ≥99.70%, Low-Metal (LM) grade semiconductor grade
Acetic acid

Transene Acetic Acid LM grade is a high-purity acetic acid suitable for semiconductor and elecrtonics applications. Low metal ion content is assured. Quart bottle.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cc
Flash Pt: 40 °C
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,3 II

Order Now

Specification Test Results

Acetic Acid 99.7 % Weight min.
Acetic Anhydride 0.01 % Weight max.
Color 7 APHA max.
Chloride 500 ppb (µmg/g) max.
Density (at 25 °C) 1,05 g/mL
Dilution Test Pass
Freezing Point 61 °F min.
Heavy Metals 500 ppb (µmg/g) max.
Solubility in Water Pass
Phosphate 500 ppb (µmg/g) max.
Residue after Evaporation 5000 ppb (µmg/g) max.
Subst. Reducing Dichromate Pass
Subst. Reducing KMnO4 Pass
Aluminum 20 ppb (µmg/g) max.
Antimony 5 ppb (µmg/g) max.
Arsenic 5 ppb (µmg/g) max.
Barium 50 ppb (µmg/g) max.
Beryllium 10 ppb (µmg/g) max.
Bismuth 50 ppb (µmg/g) max.
Boron 20 ppb (µmg/g) max.
Cadmium 10 ppb (µmg/g) max.
Calcium 100 ppb (µmg/g) max.
Chromium 10 ppb (µmg/g) max.
Cobalt 10 ppb (µmg/g) max.
Copper 20 ppb (µmg/g) max.
Gallium 10 ppb (µmg/g) max.
Germanium 20 ppb (µmg/g) max.
Gold 20 ppb (µmg/g) max.
Indium 10 ppb (µmg/g) max.
Iron 50 ppb (µmg/g) max.
Lead 20 ppb (µmg/g) max.
Lithium 10 ppb (µmg/g) max.
Magnesium 50 ppb (µmg/g) max.
Manganese 10 ppb (µmg/g) max.
Molybdenum 20 ppb (µmg/g) max.
Nickel 10 ppb (µmg/g) max.
Platinum 50 ppb (µmg/g) max.
Potassium 50 ppb (µmg/g) max.
Silver 10 ppb (µmg/g) max.
Sodium 100 ppb (µmg/g) max.
Strontium 20 ppb (µmg/g) max.
Sulfate 500 ppb (µmg/g) max.
Thallium 50 ppb (µmg/g) max.
Tin 50 ppb (µmg/g) max.
Titanium 50 ppb (µmg/g) max.
Vanadium 10 ppb (µmg/g) max.
Zinc 20 ppb (µmg/g) max.
Zirconium 50 ppb (µmg/g) max.
Particle >0.5 µm 150 pcs/mL max.
Particle >1.0 µm 10 pcs/mL max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR