Chromium etchant 1020 semiconductor grade
Supplier: Transene
|
Danger
|
0021000-GAL
0021000-QT
76541-010CS
487
USD
76541-010
76541-016
Chromium etchant 1020 semiconductor grade
Chromium etchant
Chromium Etchant 1020 is an excellent general purpose etchant for chrome thin films or photomasks.
Boiling Pt:
100 °C (water) Density: 1.16 g/cc Storage Temperature: Ambient |
MDL Number:
MFCD07370678 UN: 3264 ADR: 8,II |
Specification Test Results
Appearance | Clear orange |
Etch Capacity (rate declines at ~70%) | 65 g/gallon |
Etch Rate at 40°C | 40/sec Å/sec |
Filtration | 0,2 um |
Incompatible Materials | Al, Ni, Cu, NiCr |
pH | Acidic |
Recommended Operating Temperatures | 20-80°C (30-40°C most common) °C |
Shelf Life | 1 year |
Storage Conditions | Ambient |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...