Tetramethylammonium hydroxide 2,38% semiconductor grade NOVO DEVELOPER 342

Supplier: Transene
Danger

342-GAL 342-QT
76540-992CS 175.44 USD
76540-992 76541-052
Tetramethylammonium hydroxide 2,38% semiconductor grade NOVO DEVELOPER 342
Tetramethylammonium hydroxide

Transene NOVO Developers are available in standard concentrations for controlled developing of photoresists.

Formula: C₄H₁₃NO
MW: 91.15
Boiling Pt: 102 °C
Melting Pt: 0 °C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Order Now

Specification Test Results

TMAH Concentration 2,38 %
Typical Development Cycle
Developer 15 to 75 Seconds
Water Rinse 20 to 30 Seconds
Nitrogen Dry 15 to 20 Seconds


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR