N-Methyl-2-pyrrolidone (NMP), Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

NMP-QT NMP-GAL
76540-974EA 24 USD
76540-974 76541-022
N-Methyl-2-pyrrolidone (NMP), Low-Metal (LM) grade semiconductor grade
N-Methyl-2-pyrrolidone (NMP)

NMP is suitable for a broad range of photoresist stripping applications.

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.026 g/cc
Flash Pt: 92.78 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
Merck Index: 13,06140

Order Now

Specification Test Results

Assay 99,85 %
Color 20 APHA max.
Water 200 ppm max.
Free Amine 10 ppm max.
Aluminum 10 ppb max.
Antimony 10 ppb max.
Arsenic 50 ppb max.
Barium 10 ppb max.
Boron 10 ppb max.
Cadmium 10 ppb max.
Calcium 10 ppb max.
Chloride 0.1 ppm max.
Chromium 10 ppb max.
Cobalt 10 ppb max.
Copper 10 ppb max.
Iron 20 ppb max.
Lead 10 ppb max.
Magnesium 10 ppb max.
Manganese 10 ppb max.
Nickel 10 ppb max.
Nitrate 0.5 ppm max.
Phosphate 0.5 ppm max.
Potassium 10 ppb max.
Silver 10 ppb max.
Sodium 10 ppb max.
Sulfate 0.5 ppm max.
Tin 10 ppb max.
Titanium 10 ppb max.
Zinc 10 ppb max.
Particles (1 μm and greater) 10 max/mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR