Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade
Supplier: Transene
|
Danger
|
000BOE-QT
000BOE-GAL
76541-080EA
47
USD
76541-080
76540-945
Buffered oxide etch 10:1 (BOE 10:1) semiconductor grade
Buffered oxide etch
Buffered Oxide Etchant 10:1 is a reduced-speed etchant for oxide films. 10:1 ratio of ammonium fluoride: HF.
Boiling Pt:
103…105 °C Melting Pt: –35 °C Density: 1.11…1.12 g/cc |
UN:
2817 ADR: 8, 6.1 II |
Specification Test Results
Color | ≤ 10 APHA |
Aluminum | ≤ 3.1 ppb |
Antimony | ≤ 10 ppb |
Arsenic | ≤ 2 ppb |
Barium | ≤ 0.5 ppb |
Beryllium | ≤ 1 ppb |
Bismuth | ≤ 1 ppb |
Boron | ≤ 5 ppb |
Cadmium | ≤ 1 ppb |
Calcium | ≤ 3 ppb |
Chloride | ≤ 2 ppm |
Chromium | ≤ 2 ppb |
Cobalt | ≤ 2 ppb |
Copper | ≤ 5 ppb |
Gallium | ≤ 2 ppb |
Germanium | ≤ 2 ppb |
Gold | ≤ 2 ppb |
Iron | ≤ 5 ppb |
Lead | ≤ 2 ppb |
Lithium | ≤ 1 ppb |
Magnesium | ≤ 1 ppb |
Manganese | ≤ 2 ppb |
Molybdeum | ≤ 1 ppb |
Nickel | ≤ 1 ppb |
Nitrate | ≤ 3 ppm |
Phosphate | ≤ 0.4 ppm |
Potassium | ≤ 2 ppb |
Silver | ≤ 1 ppb |
Sodium | ≤ 5 ppb |
Strontium | ≤ 1 ppb |
Sulfate | ≤ 0.5 ppm |
Thallium | ≤ 2 ppb |
Tin | ≤ 2 ppb |
Titanium | ≤ 5 ppb |
Vanadium | ≤ 2 ppb |
Zinc | ≤ 2 ppb |
Zirconium | ≤ 2 ppb |
Particle >0.5 | ≤ 100 Particle/ml |
Particle >1.0 | ≤ 15 Particle/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...