Gold entchant TFA semiconductor grade

Supplier: Transene
Warning

015000-QT 015000-GAL
76540-930EA 123 USD
76540-930 76540-968
Gold entchant TFA semiconductor grade
Gold entchant

Gold Etchant TFA is a general purpose etchant for etching fine lines in gold thin films. Excellent photoresist compatibility. Suitable for gold bump processing. Gold Etchant TFA also etches nickel.

Order Now

Specification Test Results

Appearance Amber-brown
Etch Capacity (rate declines at ~70%) 65 g/gallon
Etch Rate at 25°C 28 Å/sec
Filtration 1 um
Incompatible Materials Ni, Cu, Al, Ag, GaAs
pH 6.5-8.0
Recommended Operating Temperatures 20-80°C (30-40°C most common) °C
Shelf Life 1 Year
Storage Conditions Ambient


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR