Focus™ 1% Acid Alcohol, StatLab
Supplier: StatLab
SL89-1
76540-468EA 46.8 USD
76540-468
Focus™ 1% Acid Alcohol, StatLab
Cleaning Agents Detergents and Surface Cleansers
Contains hydrochloric acid in 70% reagent grade alcohol. Used to remove excess stain and to produce better defined nuclei.
Available in 0.5% and 1.0% formulations.
Packaging: Manufactured in the USA