RECLAIM® Stick-On Decorative Acoustic Panels, LUXOR
Supplier: Luxor
RCLMHEX001
76539-206PK 274.25 USD
76539-206 76539-208 76539-210 76539-212 76539-214 76539-216
RECLAIM® Stick-On Decorative Acoustic Panels, LUXOR
Prefabricated Rooms
RECLAIM® Stick-on decorative acoustic panels beautify a room while helping to absorb reflection to reduce flutter echo and other common acoustic problems.
  • Available in 6-packs and 12-packs in a variety of attractive colors
  • Adhesive strips can be easily removed without damage to paint or walls*
  • Meets ASTM E84 Class A and EN13501 Class B fire standards
  • 5-year limited warranty
These 3D wall panels easily attach using included adhesive strips to help create a quieter and more inviting environment. In addition, the attractive hexagonal shape and soft colors add visual interest for enhanced aesthetics.