Cole-Parmer® Large Capacity Analog Magnetic Stirrer, Antylia Scientific
Supplier: Antyila Scientific
04807-75
76539-128EA 1127.16 USD
76539-128
Cole-Parmer® Large Capacity Analog Magnetic Stirrer, Antylia Scientific
Stirrers Magnetic Stirrers
Easy to use speed control dial.
  • Powerful magnets provide strong stirring action
This large capacity analog magnetic stirrer features power magnets and motor giving stirring speeds from 100 to 600 rpm and can mix large volumes. The stainless-steel top plate allows for a stronger magnetic coupling.