VWR® CO₂ Incubated Shaker
Supplier: VWR International
76538-838
76538-838EA 11741.19 USD
76538-838
VWR® CO₂ Incubated Shaker
Incubators CO₂ Incubators
VWR® CO₂ incubated shaker with CO₂ control.
  • CO₂ incubator with built in shaker for suspension cell culture
  • Optional universal platform for attaching flask and tube racks
  • Heated door for increased temperature uniformity
  • Stainless steel chamber with dual beam infrared sensor
The VWR® CO₂ Incubated Shaker combines the performance of CO₂ inucbators, with the added convenience of an integrated orbital shaker. The orbital shaker provides mixing up to 300 rpm and provides the optimal environment for suspension cell culture applications. All parameters (mixing speed, temperuare and CO₂ percentage) can be prgrammed on the large LED display. The incubator also inlcudes a shelf that can be used in conjuction with the shaking platform and provides still incubations.
Certifications: CE marked.