VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Supplier: Ward's
H3710-HS-GM H3710-HS-P1-GM
76538-830EA 777.4 USD
76538-830 76538-832
VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
GeneMate™ Hotplate Magnetic Stirrers, 10×10" with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • Excellent chemical resistance with white ceramic work surface
  • Stir volumes up to 15 L
  • Safety LED warns user when top plate is unsafe to touch
  • Choice of model with temperature probe for control over actual sample temperature
Certifications: CE marked.
Ordering information: 76538-832 is supplied with external temperature probe for control over actual sample temperature. Full specification details of stirrer and hotplate only models are given under separate entries, please view full details by searching for the relevant product code.