VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Supplier: Ward's
H3770-HS-GM 76538-828
76538-822EA 529.67 USD
76538-822 76538-828
VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
GeneMate™ Magnetic Hotplate Stirrers, 7×7" with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • Excellent chemical resistance with white ceramic work surface
  • Stir volumes up to 5 L
  • Safety LED warns user when top plate is unsafe to touch
  • Choice of model with temperature probe for control over actual sample temperature
Certifications: CE marked.
Ordering information: 76538-828 is supplied with external temperature probe for control over actual sample temperature. Full specification details of stirrer and hotplate only models are given under separate entries, please view full details by searching for the relevant product code.