Assay Furnaces, Model N CUP, Nabertherm
Supplier: NABERTHERM INC.
001490310-480
76538-420EA 15183.02 USD
76538-420
Assay Furnaces, Model N CUP, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These furnaces are especially used for the assay of precious metals and ashing processes where the insulation and heating must be protected from emerging gases and vapours.
  • Muffle heated from four sides (three sides for assay furnace N 25/13 CUP)
  • Heating elements and insulation protected by muffle
  • Simple replacement of muffle
  • Warm furnace can be opened
  • Tool holder on furnace
  • Stainless steel exhaust chimney above the door opening for connection of an exhaust system
  • Dual shell housing with fan cooling to reduce exterior temperatures
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
The furnace chamber forms a ceramic muffle which can easily be replaced. The assay furnace is equipped with an integrated exhaust hood above the furnace door for connection to and exhaust sytem.
Accessories information: Optional accessores include: base frame on castors (not for assay furnace N 4/13 CUP); process control and documentation via VCD software package for monitoring, documentation and control. Contact your Avantor representative for more information.
Ordering information: These assay/ashing furnaces can be ordered in a variety of sizes depending on the charge size. Models are rated up to 1280 or 1300 °C depending on the version. Control through the B500 controller (5 programs with 4 segments each).