Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm
Supplier: NABERTHERM INC.
LV155K1LN1-208 LV155K1LN1-240 LV155K1ON1-208 LV155K1ON1-240 LV035K1LN1-208 LV035K1LN1-240 LV035K1ON1-208 LV035K1ON1-240 LV055K1LN1-208 LV055K1LN1-240 LV055K1ON1-208 LV055K1ON1-240 LV095K1LN1-208 LV095K1LN1-240 LV095K1ON1-208 LV095K1ON1-240 LV035H1ON1-208 LV035H1ON1-240 LV095H1ON1-208 LV095H1ON1-240 LV155H1LN1-208 LV155H1LN1-240 LV155H1ON1-208 LV155H1ON1-240 LV035H1LN1-208 LV035H1LN1-240 LV055H1LN1-208 LV055H1LN1-240 LV055H1ON1-208 LV055H1ON1-240 LV095H1LN1-208 LV095H1LN1-240 LV034H1LN1-120 LV034H1ON1-120 LV034K1LN1-120 LV034K1ON1-120
76537-982EA 9429.71 USD
76537-982 76537-984 76537-986 76537-988 76537-990 76537-992 76537-994 76537-996 76537-998 76538-000 76538-002 76538-004 76538-006 76538-008 76538-010 76538-012 76538-014 76538-016 76538-018 76538-020 76538-022 76538-024 76538-026 76538-028 76538-030 76538-032 76538-034 76538-036 76538-038 76538-040 76538-042 76538-044 76640-510 76640-512 76640-514 76640-516
Ashing Furnaces with Fold Down or Lift Up Door, Models LV and LVT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The ashing furnaces LV 3/11 - LVT 15/11 are especially designed for ashing in the laboratory.
  • Maximum temperature: 1100 °C, heating from two sides
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fiber material, solid state relays provide for low noise operation
  • Housing made of sheets of textured stainless steel, dual shell housing for low external temperatures and stability
  • Optional fold down door (LV models) which can be used as work platform or
More Product Information