VWR® Standard Series Refrigerator and Freezer Combo Units
Supplier: VWR International
76537-060
76537-060EA 3635.78 USD
76537-060
VWR® Standard Series Refrigerator and Freezer Combo Units
Refrigerators
These multi-functional cold storage units are designed for laboratory use.
  • Mechanical thermostat temperature control
  • Spacious interior shelving
  • High density insulation
  • Auto defrost freezer
  • Two probe access ports
  • Natural, hydrocarbon refrigerant
These VWR® Combo Units offer a cold-storage solution for a variety of laboratory needs. The units provide efficient cooling with high density insulation and offer adjustable shelving, interior lighting and probe access.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency.
Material: White cabinet and door
Certifications: CSA Listed.