HyClone® Bovine Calf Serum, U.S. Origin

Supplier: Cytiva
SH30073.04 SH30073.02 SH30073.03 SH30073.02HI SH30073.03HI SH30073.04HI 3007303IR25-40 3007304IR25-40
82007-424EA 134.52 USD
82007-424 16777-204 16777-206 76237-688 76237-690 76237-692 76237-694 76237-696 76324-872 76534-626 76534-652
HyClone® Bovine Calf Serum, U.S. Origin
Sera (Serum)
HyClone Bovine Calf Serum, U.S, can be used as an effective alternative to foetal bovine serum (FBS).

  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
  • Endotoxin Concentration ≤ 10 EU/mL
  • Storage Temperature is –10 °C

Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.

Certifications: Fully traceable and conforms to all USDA regulations.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR