Cole-Parmer® Essentials Wide-Mouth Polypropylene Bottles, Antylia Scientific
Supplier: Antyila Scientific
06045-41 06045-48 06045-51 06045-56 06045-58 06045-62 06045-66 06045-68
76534-430CS 213.63 USD
76534-430 76534-432 76534-434 76534-436 76534-438 76534-440 76534-442 76534-444
Cole-Parmer® Essentials Wide-Mouth Polypropylene Bottles, Antylia Scientific
Bottles Plastic Bottles
Wide-mouth opening provides easier filling and sampling.
  • Cost-effective, rigid leakproof polypropylene (PP) bottles offer excellent chemical resistance
  • Ideal for storage and sampling of liquids or solids
  • Autoclavable
Polypropylene bottles are rigid and offer superior chemical resistance; perfect for storage and sampling of liquids and solids.
Accessories information: PP closures.