Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, PP, Antylia Scientific
Supplier: Antyila Scientific
06257-00 06257-05 06257-10 06257-20 06257-30
76534-308EA 86.91 USD
76534-308 76534-310 76534-312 76534-314 76534-316
Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, PP, Antylia Scientific
Bottles Plastic Bottles
Withstands repeated full vacuum applications.
  • Hold full vacuum for up to 24 hours
  • Polypropylene closure with thermoplastic
  • Autoclavable
These bottles can withstand repeated application of full vacuum for 24 hours at 20 °C (68 °F). The polypropylene closures feature a thermoplastic elastomer (TPE) gasket, which can be used as a waste aspirator when fitted with a filling/venting cap or barbed bulkhead fitting kit (sold separately).