Cole-Parmer® Essentials Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antyila Scientific
06028-10 06028-20 06028-30
76534-042EA 32.08 USD
76534-042 76534-044 76534-046
Cole-Parmer® Essentials Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Store chemical samples safely.
  • Leakproof polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Ideal for storing chemicals and distilled water. Larger bottles feature built-in shoulder loop.