Sentinel XL® CBRN System, with Rechargeable NIMH Battery and Duffel, ILC Dover
Supplier: ILC DOVER LP
S-5000-01
76533-628EA 2839.52 USD
76533-628
Sentinel XL® CBRN System, with Rechargeable NIMH Battery and Duffel, ILC Dover
Respirators Respiratory Systems
The Sentinel XL® CBRN PAPR system, which features an adjustable headband, is comfortable to wear and requires no fit-testing. The “lay flat” design protects the system and allows it to stow easily in a duffel bag.
  • Lightweight duffel bag included for easy transport
  • NiMH recharable battery included
  • Adjustable anti-microbial headband
  • NIOSH approved at CAP 2 rating
  • Latex-free
The Sentinel XL® CBRN PAPR system includes Butyl Hood, Sentinel XL® Blower, NiMH battery with charging cord, 3 CBRN cartridge filters, and a duffel bag.
Certifications: NIOSH approved.