Sentinel XT® Starter Kit, ILC Dover
Supplier: ILC DOVER LP
S-3327
76533-610EA 1201.45 USD
76533-610
Sentinel XT® Starter Kit, ILC Dover
Respirators Respiratory Systems
The Sentinel XT® blower, when paired with ILC Dover’s lithium polymer battery, weighs just 2.2 pounds, yet delivers 8+ CFM airflow, exceeding NIOSH requirements.
  • Proven technology with 10+ year track record of reliability
  • Brushless motor design for long life and continued quiet operation
  • Robust design with secondary impact protection chamber for air handling unit
The Sentinel XT® Starter kit includes the Sentinel XT® Blower, LiPo rechargeable battery, flow meter, charging cradle, and Quick Loc belt.