Sentinel XT® Non-Incendive Starter Kit, ILC Dover
Supplier: ILC DOVER LP
S-3326
76533-608EA 1478.8 USD
76533-608
Sentinel XT® Non-Incendive Starter Kit, ILC Dover
Respirators Respiratory Systems
The Sentinel XT® Non-incendive blower, when paired with ILC Dover’s lithium polymer battery, weighs just 2.2 pounds, yet delivers 8+ CFM airflow, exceeding NIOSH requirements.
  • Proven technology with 10+ year track record of reliability
  • Secure lock prevents battery removal without the use of a tool
  • Brushless motor design for long life and continued quiet operation
  • Robust design with secondary impact protection chamber for air handling unit
The Sentinel XT® Non-Incendive starter kit includes the Sentinel XT® Non-Incendive Blower, LiPo rechargeable battery with latch, flow meter, charging cradle, and Quick Loc belt.