Sentinel XT® Clear PAPR System with S-3112 Hood and Sentinel XT® Blower, ILC Dover
Supplier: ILC DOVER LP
S-3310
76533-598EA 1508.16 USD
76533-598
Sentinel XT® Clear PAPR System with S-3112 Hood and Sentinel XT® Blower, ILC Dover
Respirators Respiratory Systems
The Sentinel XT® PAPR system offers up to 8cfm, 33% greater airflow within the hood and suit. The hood attaches to protective clothing with clips, for a secure fit.
  • 320 degree unobstructed field of view
  • LiPo rechargeable battery lasts up to 12 hours
  • Ergonomically designed for comfort during long shifts and complex operations
  • One size fits all
  • Latex-free
The Sentinel XT® Clear PAPR System includes the S-3112 clear full hood, Sentinel XT® blower, 33" breathing tube, plug, LiPo rechargeable battery and charging cradle, Quick Loc belt, and 2 HEPA cartridge filters.