Sentinel XL® HP Clear Full Hood Conversion Kit - CBRN to XL HP Clear Full Hood with 2 HEPA Filters, ILC Dover
Supplier: ILC DOVER LP
S-3000-19
76533-556EA 317.85 USD
76533-556
Sentinel XL® HP Clear Full Hood Conversion Kit - CBRN to XL HP Clear Full Hood with 2 HEPA Filters, ILC Dover
Respirators Respiratory Systems
Convert the CBRN PAPR system into a HP PAPR System in a few easy steps.
  • Go from an APF of 1000 to a NIOSH CAP2 protection rating in just minutes
  • Save time and space, just one blower needed for CBRN and HP settings
  • Easily change out hoods and filters
  • Latex-free
The blower stays the same, only the hood and filters must be changed. Kit is supplied with 33" breathing tube, clear full hood, and 2 HEPA cartridge filters.