Sentinel XL® HP Clear Half Hood System, ILC Dover
Supplier: ILC DOVER LP
S-3000-01
76533-544EA 1563.44 USD
76533-544
Sentinel XL® HP Clear Half Hood System, ILC Dover
Respirators Respiratory Systems
Designed for use with Sentinel XL® blower, the Sentinel XL® HP clear half hood system is ideal for use in healthcare settings.
  • No fit test required
  • APF of 25
  • Can be used easily with a stethoscope
  • Available with or without duffel bag
  • Available with HiMH rechargeable battery or alkaline battery pack
  • Latex-free
Ideal for healthcare settings, the Sentinel XL® HP Clear Half Hood System includes the S-4001 clear half hood, 33" breathing tube, Sentinel XL Blower, flow meter, plug, NiMH rechargeable battery or alkaline battery pack, charging cord, Quick Loc belt, 2 HEPA filters, and carrying tote.