Sentinel XL® Blower Assembly, ILC Dover
Supplier: ILC DOVER LP
S-2002
76533-518EA 601.87 USD
76533-518
Sentinel XL® Blower Assembly, ILC Dover
Respirators Respiratory Systems
The Sentinel XL® Blower has a rugged, sealed nylon protective shell, with floating polycarbonate internal blower assembly. The dual chamber design provides enhanced durability.
  • Standard NIMH rechargeable battery pack can last up to 8 hours
  • Alkaline battery pack can last up to 10 hours
  • Blower and battery are lightweight and comfortable, weighing just 2.2 lbs.
The Sentinel XL® blower platform can be used in a variety of industries such as pharmaceutical, biopharmaceutical, infectious disease, along with chemical, biological, radiological, and nuclear applications. It can be used with Alkaline battery pack or a NiMH rechargeable battery.