VWR® Digital Rotary Evaporators
Supplier: VWR International
76533-232 76533-248 76533-250 76533-252 76533-254
76533-232EA 2712.1 USD
76533-232 76533-248 76533-250 76533-252 76533-254
VWR® Digital Rotary Evaporators
Evaporators Rotary Evaporators
LCD Digital rotary evaporators are essential instruments for use in chemical laboratories for efficient and gentle removal of solvents from samples by evaporation. Units offer an excellent distilling solution in a wide range of applications. Choice of models with automatic or manual lift.
  • Digital display shows both rotation speed and heating temperature allowing optimal control of all distillation processes
  • Automatic motor lift releases the evaporating flask to a safe position in case of power failure
  • 5 L heating bath with wide temperature range (ambient
More Product Information