Winterization Labware Kit, United Scientific Supplies
Supplier: United Scientific Supplies
WNTK2
76532-274KT 189.71 USD
76532-274
Winterization Labware Kit, United Scientific Supplies
Labware Sets
Winterization kit contains materials needed to efficiently separate compounds and refine oils.
  • Three sizes for all your needs
Each convenient package includes a filtering flask, buchner funnel, filter adapter set, and medium flow filter paper. Filtering flasks are from our FG5340 series. They are constructed using heavy wall borosilicate and include a thick rim and hose barb. The 10000 ml set includes our FFB5340 series heavy wall borosilicate filtering bottle with hose barb. Porcelain buchner funnels are from our JBF series. All kits include our FASN07 filter adapter set. Filter paper is from our FPR series and has medium porosity and flow rate. Circular filters are packaged 100 per box.
WNTK2 2000 ml Winterization Labware Kit Contents:
- (1) Filtering flask, borosilicate glass, 2000 ml (FG5340-2000)
- (1) Buchner funnel, porcelain, 2000 ml (JBF2000)
- (1) Filter adapter set of 7, synthetic rubber (FASN07)
- (1) Filter paper, medium porosity/flow, 18 cm circle, pk/100 (FPR180)
WNTK5 5000 ml Winterization Labware Kit Contents:
- (1) Filtering flask, borosilicate glass, 5000 ml (FG5340-5000)
- (1) Buchner funnel, porcelain, 4800 ml (JBF4800)
- (1) Filter adapter set of 7, synthetic rubber (FASN07)
- (1) Filter paper, medium porosity/flow, 24 cm circle, pk/100 (FPR240)
WNTK10 10000 ml Winterization Labware Kit Contents:
- (1) Filtering bottle, borosilicate glass, 10000 ml (FFB5340-10000)
- (1) Buchner funnel, porcelain, 4800 ml (JBF4800)
- (1) Filter adapter set of 7, synthetic rubber (FASN07)
- (1) Filter paper, medium porosity/flow, 24 cm circle, pk/100 (FPR240)