Volumetric Flasks, Class B, with Glass Stopper, United Scientific Supplies
Supplier: United Scientific Supplies
FG5641-10 FG5641-100 FG5641-1000 FG5641-200 FG5641-2000 FG5641-25 FG5641-250 FG5641-5 FG5641-50 FG5641-500
76531-906PK 113.28 USD
76531-906 76531-908 76531-910 76531-912 76531-914 76531-916 76531-918 76531-920 76531-922 76531-924
Volumetric Flasks, Class B, with Glass Stopper, United Scientific Supplies
Flasks Volumetric Flasks
Durable Class B Volumetric flasks.
  • Sharp graduation line and large white block lettering
Durable, machine-blown body. Heavy beaded tubing neck.
Certifications: Flasks meet Class B specifications.
Ordering information: Ground glass stoppers are included.