Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
Supplier: United Scientific Supplies
FG5639-10 FG5639-100 FG5639-1000 FG5639-20 FG5639-200 FG5639-2000 FG5639-25 FG5639-250 FG5639-5 FG5639-50 FG5639-500
76531-818EA 45.94 USD
76531-818 76531-820 76531-822 76531-830 76531-832 76531-834 76531-842 76531-844 76531-850 76531-852 76531-854
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
A ground glass stopper is included.
Certifications: Complies with ASTM E288 Class A specifications; individually serialized and certified.