Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, QR, United Scientific Supplies
Supplier: United Scientific Supplies
FG5639-1000QR FG5639-100QR FG5639-10QR FG5639-2000QR FG5639-200QR FG5639-20QR FG5639-250QR FG5639-25QR FG5639-500QR FG5639-50QR FG5639-5QR
76531-824EA 128.74 USD
76531-824 76531-826 76531-828 76531-836 76531-838 76531-840 76531-846 76531-848 76531-856 76531-858 76531-860
Volumetric Flasks, Class A, with Glass Stopper, Individually Certified, QR, United Scientific Supplies
Flasks Volumetric Flasks
Durable, machine-blown body with heavy beaded tubing neck.
  • Sharp graduation line and large white block lettering
A printed QR code allows for easy access to the calibration certificate from a mobile device or computer. A paper certificate is also included.
Certifications: Complies with ASTM E288 Class A specifications; individually serialized and certified.
Ordering information: A ground glass stopper is included.