Burette, Class B, PTFE Stopcock, United Scientific Supplies
Supplier: United Scientific Supplies
BR2130-10 BR2130-100 BR2130-25 BR2130-50
76531-490EA 35.55 USD
76531-490 76531-492 76531-494 76531-496
Burette, Class B, PTFE Stopcock, United Scientific Supplies
Burets
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
  • Amber graduations at intervals
  • Available in multiple sizes
Includes PTFE straight bore stopcock.